This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DDX50 SIRNA Assays ›
Gene Symbol:
Ddx50
Gene Name:
DEAD (Asp-Glu-Ala-Asp) box polypeptide 50
Gene Aliases:
4933429B04Rik, 8430408E17Rik, GU2, RH-II
Chromosome Location:
Chr.10: 62616015 - 62651606 on Build GRCm38
UniGene ID:
Mm.114116
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s97029

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ddx50
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053183.2 NP_444413.1 12 1815
GenBank mRNA AF334104.1 - 12 1816
AK030239.1 - 13 1887
AK041281.1 - 12 1714
AK162347.1 - 3 2190
AK213799.1 - 1 15
BC049906.1 - 12 1816
BC055440.1 - 12 1810
BC075714.1 - 12 1768
BC090996.1 - 12 1835 6757408

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
RNA secondary structure unwinding
nucleotide binding
nucleic acid binding
RNA binding
ATP-dependent RNA helicase activity
helicase activity
ATP binding
hydrolase activity
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header