This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KLRA22 SIRNA Assays ›
Gene Symbol:
Klra22 Klra1
Gene Name:
killer cell lectin-like receptor subfamily A, member 22
killer cell lectin-like receptor, subfamily A, member 1
Gene Aliases:
Ly49v
A1, CH29-493D4.3, Klra22, Ly49a, Ly49o<129>, Ly49v
Chromosome Location:
-
Chromosome Location:
Chr.6: 130363918 - 130386874 on Build GRCm38
UniGene ID:
Mm.333431
Mm.24399
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s96905

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Klra22
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053152.2 NP_444382.1 4 626
GenBank mRNA AF288381.1 - 4 626
Gene Symbol: Klra1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF419250.1 - 6 965
AF444273.1 - 6 965

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0