This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other WDR6 SIRNA Assays ›
Gene Symbol:
Wdr6
Gene Name:
WD repeat domain 6
Gene Aliases:
mWDR6
Chromosome Location:
Chr.9: 108572313 - 108578670 on Build GRCm38
UniGene ID:
Mm.335454
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s96413

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Wdr6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031392.2 NP_113569.1 2 1507
GenBank mRNA AB041854.1 - 2 1576
AF348591.1 - 2 1503
AK037181.1 - 2 1484
AK046119.1 - 2 1474
AK052235.1 - 2 1448
AK146617.1 - 2 1464
AK163752.1 - 2 1440
AK165935.1 - 2 1471
AK166265.1 - 2 1484
AK169357.1 - 2 1508
AK199272.1 - 1 153
AK203715.1 - 1 153
BC050894.1 - 2 1507 5708880
BC054367.1 - 2 1435 5346943

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell cycle
cell cycle arrest
negative regulation of cell proliferation
negative regulation of autophagy
insulin receptor substrate binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0