This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRIM2 SIRNA Assays ›
Gene Symbol:
Trim2
Gene Name:
tripartite motif-containing 2
Gene Aliases:
mKIAA0517, narf
Chromosome Location:
Chr.3: 84160439 - 84305832 on Build GRCm38
UniGene ID:
Mm.44876
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s96101

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trim2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271725.1 NP_001258654.1 4 710
NM_001271726.1 NP_001258655.1 4 746
NM_001271727.1 NP_001258656.1 4 1015
NM_001271728.1 NP_001258657.1 4 813
NM_030706.3 NP_109631.2 4 809
GenBank mRNA AB043550.1 - 4 793
AF220017.1 - 4 658
AK039741.1 - 4 807
AK045410.1 - 4 806
AK079415.1 - 4 960
AK141419.1 - 4 656
AK147275.1 - 4 923
AK147395.1 - 4 804
AK164350.1 - 3 512
AK220343.1 - 4 1014
BC058668.1 - 4 777
BC058961.1 - 4 813 6826143

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
regulation of neuron apoptotic process
ubiquitin-protein transferase activity
protein binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
myosin binding
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0