This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CWC22 SIRNA Assays ›
Gene Symbol:
Cwc22 Gm13698
Gene Name:
CWC22 spliceosome-associated protein homolog (S. cerevisiae)
CWC22 spliceosome-associated protein homolog pseudogene
Gene Aliases:
AA684037, AI173004, AL022752, B230213M24, mKIAA1604
OTTMUSG00000013395
Chromosome Location:
Chr.2: 77895653 - 77946356 on Build GRCm38
Chromosome Location:
Chr.2: 83955605 - 83958658 on Build GRCm38
UniGene ID:
Mm.288151
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s96020

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cwc22
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290740.1 NP_001277669.1 20 2965
NM_030560.5 NP_085037.2 21 3041
NM_172667.2 NP_766255.1 20 2962
GenBank mRNA AK045589.1 - 20 2794
AK077961.1 - 21 2869
AK129404.1 - 4264
AK132001.1 - 20 2906
BC003273.1 - 13 1907
BC003993.1 - 20 2778 3492146
Gene Symbol: Gm13698
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK139594.1 - 2 823

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0