This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DESI2 SIRNA Assays ›
Gene Symbol:
Desi2
Gene Name:
desumoylating isopeptidase 2
Gene Aliases:
5830417C01Rik, DeSI-2, Fam152a, Pppde1
Chromosome Location:
Chr.1: 178187231 - 178252598 on Build GRCm38
UniGene ID:
Mm.440132
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s95743

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Desi2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024282.3 NP_077244.1 4 761
GenBank mRNA AK020014.1 - 4 646
AK029574.1 - 5 835
AK049579.1 - 4 530
AK146492.1 - 4 494
AK160733.1 - 4 350
AK216786.1 - 3 233
BC002200.1 - 4 430 3489351
BC046816.1 - 4 468 5702476

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header