This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRAPPC6B SIRNA Assays ›
Gene Symbol:
Trappc6b
Gene Name:
trafficking protein particle complex 6B
Gene Aliases:
5830498C14Rik, C79212
Chromosome Location:
Chr.12: 59043092 - 59061489 on Build GRCm38
UniGene ID:
Mm.290070
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s95389

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trappc6b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK020026.1 - 3 383
AK046610.1 - 3 411
AK089083.1 - 3 224
AK168294.1 - 4 671
BC031464.1 - 3 368 4017482

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106733, FQ230024.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0