siRNA Details
Gene Symbol: Heg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175256.5 | NP_780465.4 | 6, 7 | 2443 | ||
GenBank mRNA | BC076596.1 | - | 4, 5 | 1722 | 30645937 |

Gene Symbol: Heg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175256.5 | NP_780465.4 | 6, 7 | 2443 | ||
GenBank mRNA | BC076596.1 | - | 4, 5 | 1722 | 30645937 |