This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CTRC SIRNA Assays ›
Gene Symbol:
Ctrc
Gene Name:
chymotrypsin C (caldecrin)
Gene Aliases:
1810044E12Rik
Chromosome Location:
Chr.4: 141838239 - 141846359 on Build GRCm38
UniGene ID:
Mm.489680
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s94575

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ctrc
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033875.2 NP_001029047.1 7 801
GenBank mRNA AK007772.1 - 6 629
BC103715.1 - 7 799 6430373
BC115516.1 - 7 801 40101127
BC115517.1 - 7 801 40101128

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
cellular calcium ion homeostasis
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0