siRNA Details
Gene Symbol: 4930563E22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163728.1 | NP_001157200.1 | 1 | 1226 | ||
GenBank mRNA | AK044443.1 | - | 1 | 1227 | ||
AK132388.1 | - | 1 | 1179 |

Gene Symbol: 4930563E22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163728.1 | NP_001157200.1 | 1 | 1226 | ||
GenBank mRNA | AK044443.1 | - | 1 | 1227 | ||
AK132388.1 | - | 1 | 1179 |