This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM63A SIRNA Assays ›
Gene Symbol:
Fam63a
Gene Name:
family with sequence similarity 63, member A
Gene Aliases:
1810005H09Rik, 4930504E06Rik, AA562754, AA960384, AW561904, cI-40, mKIAA1390, ni
Chromosome Location:
Chr.3: 95281590 - 95296164 on Build GRCm38
UniGene ID:
Mm.279287
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s93216

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam63a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133858.4 NP_598619.2 8 2038
NM_199475.1 NP_955769.1 8 2038
GenBank mRNA AB075603.1 - 7 1049
AK036838.1 - 8 2030
AK087859.1 - 8 1853
AK169673.1 - 8 2038
AK182185.1 - 3 305
AK206919.1 - 3 305
AK215644.1 - 3 305
AK219539.1 - 2 173
BC039762.1 - 7 856
BC051048.1 - 7 921
BC057901.1 - 6 567
BC069953.1 - 1 42
BC093521.1 - 4 607 4007022
BC094388.1 - 8 1855 5708548
BC132301.1 - 7 1283 40130578
BC144749.1 - 7 1283 9053297

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0