siRNA Details
Gene Symbol: Kbtbd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028974.1 | NP_083250.1 | 1 | 958 | ||
GenBank mRNA | AK048611.1 | - | 1 | 851 | ||
BC139793.1 | - | 2 | 838 |

Gene Symbol: Kbtbd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028974.1 | NP_083250.1 | 1 | 958 | ||
GenBank mRNA | AK048611.1 | - | 1 | 851 | ||
BC139793.1 | - | 2 | 838 |