This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TSPAN17 SIRNA Assays ›
Gene Symbol:
Tspan17
Gene Name:
tetraspanin 17
Gene Aliases:
2210021G21Rik, AI047581, Fbxo23, Tm4sf17
Chromosome Location:
Chr.13: 54789352 - 54796896 on Build GRCm38
UniGene ID:
Mm.32883
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s92570

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tspan17
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028841.3 NP_083117.2 4 552
GenBank mRNA AI019499.1 - 1 23
AK008761.1 - 4 497
AK166272.1 - 4 555
AK171691.1 - 4 501
BC010346.1 - 4 497 2811935

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001013138, XM_006253630, XM_006253631, AY485933, BC097431.

Gene Ontology Categories:

Function(s) Process(es)

protein maturation
establishment of protein localization to organelle
enzyme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0