This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FBXL22 SIRNA Assays ›
Gene Symbol:
Fbxl22
Gene Name:
F-box and leucine-rich repeat protein 22
Gene Aliases:
1110004B15Rik
Chromosome Location:
Chr.9: 66508459 - 66514593 on Build GRCm38
UniGene ID:
Mm.297998
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s92393

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fbxl22
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175206.4 NP_780415.1 2 1110
GenBank mRNA AK052175.1 - 2 1096
AK084845.1 - 2 1027
AK085980.1 - 2 1053

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0