This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CROT SIRNA Assays ›
Gene Symbol:
Crot
Gene Name:
carnitine O-octanoyltransferase
Gene Aliases:
1200003H03Rik
Chromosome Location:
Chr.5: 8966023 - 8997146 on Build GRCm38
UniGene ID:
Mm.28197
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s92288

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Crot
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023733.3 NP_076222.1 4 386
GenBank mRNA AK004567.2 - 4 387
AK037973.1 - 3 838
BC006593.1 - 4 374 3156501
BC012308.1 - 4 401 3500167

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

generation of precursor metabolites and energy
lipid metabolic process
fatty acid metabolic process
fatty acid beta-oxidation
transport
carnitine metabolic process
fatty acid transport
coenzyme A metabolic process
response to drug
medium-chain fatty acid metabolic process
receptor binding
carnitine O-octanoyltransferase activity
transferase activity
transferase activity, transferring acyl groups

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header