This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other USP16 SIRNA Assays ›
Gene Symbol:
Usp16
Gene Name:
ubiquitin specific peptidase 16
Gene Aliases:
1200004E02Rik, 2810483I07Rik, 6330514E22Rik, UBPM
Chromosome Location:
Chr.16: 87454708 - 87483515 on Build GRCm38
UniGene ID:
Mm.152941
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s92284

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Usp16
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024258.2 NP_077220.2 15 2188
GenBank mRNA AK004594.2 - 15 2163
AK013437.1 - 9 1204
AK160038.1 - 15 2158
BC003278.1 - 15 2117 3582180
BC004577.1 - 3 700

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
proteolysis
ubiquitin-dependent protein catabolic process
cellular response to DNA damage stimulus
cell cycle
mitotic nuclear division
chromatin modification
histone deubiquitination
monoubiquitinated histone H2A deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of transcription, DNA-templated
positive regulation of translational elongation
positive regulation of transcription from RNA polymerase II promoter
protein homotetramerization
cell division
regulation of proteasomal protein catabolic process
histone H2A K63-linked deubiquitination
transcription coactivator activity
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
protein binding
peptidase activity
cysteine-type peptidase activity
zinc ion binding
hydrolase activity
thiol-dependent ubiquitinyl hydrolase activity
histone binding
ubiquitin binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0