This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CEP55 SIRNA Assays ›
Gene Symbol:
Cep55
Gene Name:
centrosomal protein 55
Gene Aliases:
1200008O12Rik, 2700032M20Rik
Chromosome Location:
Chr.19: 38054981 - 38074425 on Build GRCm38
UniGene ID:
Mm.9916
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s92275

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cep55
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164362.1 NP_001157834.1 9 1521
NM_028293.1 NP_082569.1 8 1145
NM_028760.2 NP_083036.2 9 1497
GenBank mRNA AK004655.1 - 9 1523
AK028216.1 - 8 1131
AK088548.1 - 9 1204
BC026966.1 - 6 853
BC031396.1 - 9 1450 4950530

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001025646, XM_006231318, XM_006231320, XM_006231319, BC098013.

Gene Ontology Categories:

Function(s) Process(es)

mitotic cytokinesis
cell separation after cytokinesis
cell cycle
mitotic nuclear division
establishment of protein localization
cell division
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0