This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ASZ1 SIRNA Assays ›
Gene Symbol:
Asz1
Gene Name:
ankyrin repeat, SAM and basic leucine zipper domain containing 1
Gene Aliases:
4933400N19Rik, Gasz, ORF3
Chromosome Location:
Chr.6: 18050964 - 18109061 on Build GRCm38
UniGene ID:
Mm.73091
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s92223

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Asz1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023729.3 NP_076218.3 5 567
GenBank mRNA AF459789.1 - 5 561
AK016595.1 - 5 562
BC058859.1 - 5 576 30011651
BG088136.2 - 5 567

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_130750, AF461260.

Gene Ontology Categories:

Function(s) Process(es)

male meiosis
signal transduction
multicellular organism development
germ cell development
spermatogenesis
cell differentiation
gene silencing by RNA
piRNA metabolic process
DNA methylation involved in gamete generation
meiotic cell cycle
signal transducer activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0