This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM161A SIRNA Assays ›
Gene Symbol:
Fam161a
Gene Name:
family with sequence similarity 161, member A
Gene Aliases:
4930430E16Rik
Chromosome Location:
Chr.11: 23007317 - 23045743 on Build GRCm38
UniGene ID:
Mm.252406
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s92075

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam161a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028672.2 NP_082948.2 3 756
GenBank mRNA AK133447.1 - 3 772
BC024460.1 - 4 689 4975427
BC026495.1 - 3 756 4505506

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell projection organization
cilium assembly
cilium organization
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0