This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 1700080E11RIK SIRNA Assays ›
Gene Symbol:
1700080E11Rik
Gene Name:
RIKEN cDNA 1700080E11 gene
Gene Aliases:
AI429110, Tseg8
Chromosome Location:
Chr.9: 105143344 - 105145082 on Build GRCm38
UniGene ID:
Mm.23516
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s91822

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1700080E11Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028562.3 NP_082838.2 2 431
GenBank mRNA AK006953.1 - 2 432
BC048436.1 - 2 420 6773931
FJ211595.1 - 2 277

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0