This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HSPA12A SIRNA Assays ›
Gene Symbol:
Hspa12a
Gene Name:
heat shock protein 12A
Gene Aliases:
1700063D12Rik, AI118035, AI840429, AW048913, AW556406, D5Mgi40, mKIAA0417
Chromosome Location:
Chr.19: 58795750 - 58943545 on Build GRCm38
UniGene ID:
Mm.39739
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s91770

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hspa12a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175199.3 NP_780408.1 10 1163
GenBank mRNA AB093239.1 - 10 1107
AK038491.1 - 10 1135
AK087732.1 - 11 1321
AK141925.1 - 10 1164
AY196789.1 - 10 1162
BC030362.1 - 10 1133 5369051

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
nucleotide binding
molecular_function
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0