This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP518A SIRNA Assays ›
Gene Symbol:
Zfp518a
Gene Name:
zinc finger protein 518A
Gene Aliases:
2810401C22Rik, 6330417C12Rik, AI426493, Zfp518, Znf518, Znf518a, mKIAA0335
Chromosome Location:
Chr.19: 40893930 - 40917947 on Build GRCm38
UniGene ID:
Mm.313523
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s91206

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp518a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028319.1 NP_082595.1 5 2199
GenBank mRNA BC138382.1 - 3 1905 8861402
BC138383.1 - 3 1905 8861403

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0