This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNHG5 SIRNA Assays ›
Gene Symbol:
Snhg5 9330159M07Rik 9430037G07Rik
Gene Name:
small nucleolar RNA host gene 5
RIKEN cDNA 9330159M07 gene
RIKEN cDNA 9430037G07 gene
Gene Aliases:
2810026P18Rik, 4933428I09Rik, mU50HG-b
1700096K11Rik, mU50hg-a
-
Chromosome Location:
Chr.9: 88521052 - 88522897 on Build GRCm38
Chromosome Location:
Chr.9: 88841798 - 88858870 on Build GRCm38
Chromosome Location:
Chr.9: 88595325 - 88599243 on Build GRCm38
UniGene ID:
Mm.21798
Mm.149040
Mm.427378
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
siRNA ID s91185

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Snhg5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK012825.1 - 5 806
BC032970.1 - 5 789
KF876690.1 - 5 808
Gene Symbol: 9330159M07Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK136619.1 - 5 420
Gene Symbol: 9430037G07Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK034782.1 - 3 1027
AK050253.1 - 5 466

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0