This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LIN9 SIRNA Assays ›
Gene Symbol:
Lin9
Gene Name:
lin-9 homolog (C. elegans)
Gene Aliases:
2700022J23Rik, Bara, Lin-9, TGS, TGS1, mLin-9
Chromosome Location:
Chr.1: 180641150 - 180690687 on Build GRCm38
UniGene ID:
Mm.275044
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s91114

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lin9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001103182.2 NP_001096652.2 5 534
GenBank mRNA AF190325.1 - 5 350
AK042357.1 - 5 316
AK052618.1 - 5 332
AK089989.1 - 5 535
BC069181.1 - 2 73
BC088992.1 - 5 324
BC099695.1 - 5 318

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_173083, NM_001270409, NM_001270410, AY166858, AF190324, and others.

Gene Ontology Categories:

Function(s)

transcription, DNA-templated
cell cycle
DNA biosynthetic process

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0