This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM87B SIRNA Assays ›
Gene Symbol:
Tmem87b
Gene Name:
transmembrane protein 87B
Gene Aliases:
2610301K12Rik, 2810431I02Rik, AU014804
Chromosome Location:
Chr.2: 128818072 - 128854261 on Build GRCm38
UniGene ID:
Mm.259688
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s91021

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem87b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028248.2 NP_082524.2 4 425
GenBank mRNA AK011959.1 - 4 346
AK050203.1 - 4 417
AK164504.1 - 4 407
AK166991.1 - 4 405
AK171811.1 - 5 341
AK172611.1 - 5 341
BC031488.1 - 4 381
CA479832.1 - 4 647

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191854, XM_006234976, XM_008762182.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header