This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KATNBL1 SIRNA Assays ›
Gene Symbol:
Katnbl1 Slc12a6
Gene Name:
katanin p80 subunit B like 1
solute carrier family 12, member 6
Gene Aliases:
2410042D21Rik, AA166487, AA473307, AI467442
9530023I19Rik, KCC3, gaxp
Chromosome Location:
Chr.2: 112377107 - 112414645 on Build GRCm38
Chromosome Location:
Chr.2: 112265538 - 112363163 on Build GRCm38
UniGene ID:
Mm.425091
Mm.491155
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s90974

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Katnbl1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024254.3 NP_077216.1 2 202
GenBank mRNA AK010654.1 - 2 181
AK036861.1 - 2 202
AK049599.1 - 2 188
AK151464.1 - 2 188
AK152570.1 - 2 167
AK162183.1 - 2 132
AK168639.1 - 2 200
BC003216.1 - 2 158 3157982
BC012402.1 - 2 158 3157982
BC080661.1 - 2 158 4460520
Gene Symbol: Slc12a6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC062099.1 - 4184 6844732

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109645, XM_006234712, BC161951.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0