This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VPS9D1 SIRNA Assays ›
Gene Symbol:
Vps9d1
Gene Name:
VPS9 domain containing 1
Gene Aliases:
1300018I17Rik, 2410004N05Rik, 5DOS1, Vsp9d1
Chromosome Location:
Chr.8: 123242356 - 123254217 on Build GRCm38
UniGene ID:
Mm.276193
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s90882

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vps9d1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028200.1 NP_082476.1 9 887
GenBank mRNA AK005046.2 - 9 839
AK010402.1 - 9 820
AK028751.1 - 9 830
AK032561.1 - 8 1344
AK151498.1 - 9 893
AK154409.1 - 9 805
AK155433.1 - 9 805
AY751521.1 - 9 1060
BC057621.1 - 9 888 6402384

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header