This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MMS19 SIRNA Assays ›
Gene Symbol:
Mms19
Gene Name:
MMS19 (MET18 S. cerevisiae)
Gene Aliases:
2410001K24Rik, 2610042O15Rik, AI316855, C79368, C86341, Mms19l
Chromosome Location:
Chr.19: 41943707 - 41981159 on Build GRCm38
UniGene ID:
Mm.218940
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s90748

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mms19
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028152.3 NP_082428.1 6 492
GenBank mRNA AF319949.1 - 6 486
AK010326.1 - 6 496
AK011754.1 - 6 486
AK028317.1 - 6 492
AK030619.1 - 6 493
AK036983.1 - 6 486
AK044710.1 - 1 1191
AK050265.1 - 2 807
AK078688.1 - 6 485
AK087633.1 - 6 493
BC018614.1 - 6 470
BC050817.1 - 6 472 6332214

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA metabolic process
DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
chromosome segregation
iron-sulfur cluster assembly
positive regulation of transcription, DNA-templated
transcription coactivator activity
estrogen receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0