This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PGM2 SIRNA Assays ›
Gene Symbol:
Pgm2
Gene Name:
phosphoglucomutase 2
Gene Aliases:
2610020G18Rik, AA407108, AI098105, Pgm-2, Pgm1
Chromosome Location:
Chr.4: 99929451 - 99987294 on Build GRCm38
UniGene ID:
Mm.217764
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s90686

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pgm2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028132.3 NP_082408.3 8, 9 1388
GenBank mRNA AK011485.1 - 8, 9 1313
AK147982.1 - 8, 9 1360
AK178518.1 - 1, 2 61
AK186198.1 - 2, 3 257
AK187718.1 - 2, 3 247
AK189464.1 - 1, 2 61
BC008527.1 - 8, 9 1332 3495694
BC036290.1 - 8, 9 1576
BC055713.1 - 8, 9 1339
BC066828.1 - 1 400
BC080801.1 - 8, 9 1359

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
glycogen biosynthetic process
glucose metabolic process
galactose catabolic process
organic substance metabolic process
magnesium ion binding
phosphoglucomutase activity
isomerase activity
intramolecular transferase activity, phosphotransferases
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0