This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC38A10 SIRNA Assays ›
Gene Symbol:
Slc38a10
Gene Name:
solute carrier family 38, member 10
Gene Aliases:
1810073N04Rik
Chromosome Location:
Chr.11: 120103951 - 120151351 on Build GRCm38
UniGene ID:
Mm.253403
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s90535

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc38a10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164798.1 NP_001158270.1 17 3242
NM_001164799.1 NP_001158271.1 16 3221
NM_001164800.1 NP_001158272.1 16 3218
NM_024249.5 NP_077211.4 17 3245
GenBank mRNA AK032245.1 - 17 3169
AK159948.1 - 16 3183
AK165672.1 - 16 3185
BC005641.1 - 5 1594
BC023375.1 - 1 374
BC043299.1 - 7 1918
BC055017.1 - 4 1266
BC082300.1 - 16 3189 30362204
BC088811.1 - 17 3178 6493546

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
ion transport
sodium ion transport
amino acid transport
bone development
molecular_function
amino acid transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0