This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZSWIM2 SIRNA Assays ›
Gene Symbol:
Zswim2
Gene Name:
zinc finger SWIM-type containing 2
Gene Aliases:
1700025P14Rik, 4933437F18Rik, MEX
Chromosome Location:
Chr.2: 83915079 - 83941254 on Build GRCm38
UniGene ID:
Mm.116957
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s90187

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zswim2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027964.2 NP_082240.1 9 1436
GenBank mRNA AK006367.1 - 9 1437
BC120837.1 - 9 1416 40129760
BC144860.1 - 9 1404
EU233996.1 - 7 1551

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
apoptotic process
positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
ubiquitin-protein transferase activity
protein binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
protein self-association
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0