This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TELO2 SIRNA Assays ›
Gene Symbol:
Telo2
Gene Name:
TEL2, telomere maintenance 2, homolog (S. cerevisiae)
Gene Aliases:
1200003M09Rik, AI415602, Tel2, mKIAA0683
Chromosome Location:
Chr.17: 25099569 - 25115971 on Build GRCm38
UniGene ID:
Mm.288702
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s89924

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Telo2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163661.1 NP_001157133.1 16 2519
NM_027880.2 NP_082156.2 16 2430
GenBank mRNA AK004582.1 - 16 2300
AK028658.1 - 16 2284
AK045321.1 - 9 1966
AK050773.1 - 16 2249
AK129189.1 - 15 2592
AK143827.1 - 16 2520
AK146963.1 - 16 2306
AK154727.1 - 16 2332
BC011077.1 - 16 2280 3984911
BC080713.1 - 15 2539

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of TOR signaling
protein complex binding
protein complex scaffold

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0