This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MCM9 SIRNA Assays ›
Gene Symbol:
Mcm9
Gene Name:
minichromosome maintenance complex component 9
Gene Aliases:
9030408O17Rik, Gm235, Mcmdc1
Chromosome Location:
Chr.10: 53536315 - 53630439 on Build GRCm38
UniGene ID:
Mm.278221
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s89773

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mcm9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027830.2 NP_082106.2 14 3821
GenBank mRNA AK133212.1 - 7 2123
AK133345.1 - 7 2936
BC062185.1 - 5 1889

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
DNA replication
DNA repair
cellular response to DNA damage stimulus
gamete generation
female gamete generation
nucleotide binding
DNA binding
helicase activity
protein binding
ATP binding
hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0