siRNA Details
Gene Symbol: Tprg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175165.3 | NP_780374.1 | 2 | 256 | ||
GenBank mRNA | AK017323.1 | - | 2 | 252 | ||
AK036796.1 | - | 2 | 259 |

Gene Symbol: Tprg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175165.3 | NP_780374.1 | 2 | 256 | ||
GenBank mRNA | AK017323.1 | - | 2 | 252 | ||
AK036796.1 | - | 2 | 259 |