This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other QPCT SIRNA Assays ›
Gene Symbol:
Qpct
Gene Name:
glutaminyl-peptide cyclotransferase (glutaminyl cyclase)
Gene Aliases:
5730422A13Rik
Chromosome Location:
Chr.17: 79051906 - 79090268 on Build GRCm38
UniGene ID:
Mm.293870
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s88921

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Qpct
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027455.2 NP_081731.1 3 723
GenBank mRNA AK017598.1 - 3 386
AK037197.1 - 3 376
AK045974.1 - 3 724
AK148394.1 - 3 394
BC020023.1 - 2 294 3491756
BC151027.1 - 3 467 9087940
BC151029.1 - 3 467 9087942

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001134557, XM_006239632, FQ228724, FQ229106.

Gene Ontology Categories:

Function(s) Process(es)

peptidyl-pyroglutamic acid biosynthetic process, using glutaminyl-peptide cyclotransferase
zinc ion binding
glutaminyl-peptide cyclotransferase activity
transferase activity
transferase activity, transferring acyl groups
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0