This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RUFY2 SIRNA Assays ›
Gene Symbol:
Rufy2
Gene Name:
RUN and FYVE domain-containing 2
Gene Aliases:
2610111M19Rik, AI844453, Denn, LZ-FYVE, ZFYVE13
Chromosome Location:
Chr.10: 62980223 - 63018742 on Build GRCm38
UniGene ID:
Mm.234587
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s88833

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rufy2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027425.3 NP_081701.2 9 838
GenBank mRNA AK011847.1 - 9 808
AK083205.1 - 9 823
AK173195.1 - 9 799
BC017645.1 - 9 800
BC038265.1 - 8 709
BC137736.1 - 9 787 8860755
BC137737.1 - 9 787 8860756

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001168586, XM_006256393, XM_008772948, XM_008772927, XM_008772951, and others.

Gene Ontology Categories:

Process(es)

metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0