This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UBE2Q1 SIRNA Assays ›
Gene Symbol:
Ube2q1
Gene Name:
ubiquitin-conjugating enzyme E2Q (putative) 1
Gene Aliases:
1110002C01Rik, 2310012M18Rik, AI854014, Gtap, NICE-5, PRO3094, Ube2q
Chromosome Location:
Chr.3: 89773609 - 89783997 on Build GRCm38
UniGene ID:
Mm.41438
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s88537

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ube2q1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027315.4 NP_081591.3 5 819
GenBank mRNA AK009324.1 - 4 401
AK146707.1 - 5 632
AK177704.1 - 4 371
AK214106.1 - 4 338
AY112699.1 - 5 812
AY945937.1 - 5 716
BC030044.1 - 2 144
BC060966.1 - 3 212
BC082275.1 - 5 642 6310111

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_017582, BC148185, AY112698, AK293986, BC087836, and others.
This siRNA also targets the following transcripts in Rat: NM_001106448, BC166534.

Gene Ontology Categories:

Function(s) Process(es)

suckling behavior
embryo implantation
mating behavior
fertilization
protein ubiquitination
reproductive system development
prolactin secretion
nucleotide binding
ATP binding
ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0