This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TTC39B SIRNA Assays ›
Gene Symbol:
Ttc39b
Gene Name:
tetratricopeptide repeat domain 39B
Gene Aliases:
1810054D07Rik, 9130422G05Rik, AI450603, AV103077, AW215585
Chromosome Location:
Chr.4: 83220301 - 83324270 on Build GRCm38
UniGene ID:
Mm.5540
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s88318

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ttc39b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027238.2 NP_081514.1 8 677
GenBank mRNA AK037208.1 - 8 678
AK049108.1 - 8 728
AK132655.1 - 3 128
BC151021.1 - 8 639 9087934

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001168341, NM_001168339, NM_001168342, NM_001168340, NM_152574, and others.
This siRNA also targets the following transcripts in Rat: NM_001106665, XM_006238350, XM_008763774, XM_008763773.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header