This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MTERF4 SIRNA Assays ›
Gene Symbol:
Mterf4
Gene Name:
mitochondrial transcription termination factor 4
Gene Aliases:
1810059A23Rik, 4933412H03Rik, Mterfd2
Chromosome Location:
Chr.1: 93299211 - 93305965 on Build GRCm38
UniGene ID:
Mm.52809
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s88274

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mterf4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178051.3 NP_835152.1 2 400
GenBank mRNA AK007901.1 - 2 387
AK033043.1 - 2 411
AK077142.1 - 2 400
AK079098.1 - 2 392
BC002280.1 - 1 13 3497421
BC055877.1 - 1 344 4236729
BC064709.1 - 2 393 30314353
BY261356.1 - 2 406

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
rRNA processing
transcription from mitochondrial promoter
protein targeting to mitochondrion
heart development
ribosome assembly
camera-type eye development
double-stranded DNA binding
RNA binding
rRNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0