This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DYRK2 SIRNA Assays ›
Gene Symbol:
Dyrk2
Gene Name:
dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2
Gene Aliases:
1810038L18Rik
Chromosome Location:
Chr.10: 118855602 - 118889246 on Build GRCm38
UniGene ID:
Mm.45565
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s87547

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dyrk2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014390.2 NP_001014412.1 3 1186
GenBank mRNA AK149371.1 - 3 1422
AK155496.1 - 3 1436
BC085145.1 - 3 1186 6808145

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_006482, NM_003583, Y13493, BC005809, AK313937, and others.
This siRNA also targets the following transcripts in Rat: NM_001108100, XM_006241390, XM_006241389, XM_008765401, XM_006241391.

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
apoptotic process
smoothened signaling pathway
phosphorylation
peptidyl-tyrosine phosphorylation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
positive regulation of glycogen biosynthetic process
negative regulation of NFAT protein import into nucleus
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
protein serine/threonine/tyrosine kinase activity
protein tyrosine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
manganese ion binding
ubiquitin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0