siRNA Details
Gene Symbol: Ubr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160319.1 | NP_001153791.1 | 48 | 7040 | ||
GenBank mRNA | AB093242.2 | - | 21 | 3147 |

Gene Symbol: Ubr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160319.1 | NP_001153791.1 | 48 | 7040 | ||
GenBank mRNA | AB093242.2 | - | 21 | 3147 |