This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SCGB2A2 SIRNA Assays ›
Gene Symbol:
SCGB2A2
Gene Name:
secretoglobin family 2A member 2
Gene Aliases:
MGB1, UGB2
Chromosome Location:
Chr.11: 62270130 - 62273160 on Build GRCh38
UniGene ID:
Hs.46452
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s8742

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SCGB2A2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002411.3 NP_002402.1 2 280
XM_005274005.3 XP_005274062.3 2 305
GenBank mRNA AI904278.1 - 2 280
AY217100.1 - 2 218
BC067220.1 - 2 281 30321681
BC128252.1 - 2 272 40115103
BC128402.1 - 2 272 40115099
U33147.1 - 2 278

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0