This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZDHHC3 SIRNA Assays ›
Gene Symbol:
Zdhhc3
Gene Name:
zinc finger, DHHC domain containing 3
Gene Aliases:
1110020O22Rik, 1810006O10Rik, 2210017C02Rik, DHHC-3, GODZ, Gramp1, Zfp373
Chromosome Location:
Chr.9: 123066802 - 123113205 on Build GRCm38
UniGene ID:
Mm.28300
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s87362

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zdhhc3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026917.5 NP_081193.2 7 1029
GenBank mRNA AB088044.1 - 6 795
AK007355.1 - 8 1501
AK018755.1 - 7 996
AK080541.1 - 7 995
AY166672.1 - 6 795
BC005531.1 - 8 1056
BC018334.1 - 6 865
BC025126.1 - 7 1014 3669723
BC052464.1 - 7 934 6834763

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001135179, NM_016598, BC015467.

Gene Ontology Categories:

Function(s) Process(es)

protein targeting
protein palmitoylation
zinc ion binding
palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
protein-cysteine S-palmitoyltransferase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0