This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DUSP23 SIRNA Assays ›
Gene Symbol:
Dusp23
Gene Name:
dual specificity phosphatase 23
Gene Aliases:
1300005N15Rik, LDP-3
Chromosome Location:
Chr.1: 172630769 - 172632974 on Build GRCm38
UniGene ID:
Mm.33024
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s86665

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dusp23
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026725.2 NP_081001.1 2 490
GenBank mRNA AB183013.1 - 2 484
AK004912.1 - 2 381
BC069187.1 - 2 484 850475

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
dephosphorylation
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
hydrolase activity
phosphatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0