This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GSTM7 SIRNA Assays ›
Gene Symbol:
Gstm7
Gene Name:
glutathione S-transferase, mu 7
Gene Aliases:
0610005A07Rik, AI894236, Cd203c
Chromosome Location:
Chr.3: 107926332 - 107931873 on Build GRCm38
UniGene ID:
Mm.458189
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s86554

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gstm7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026672.2 NP_080948.2 7 567
GenBank mRNA AK002213.1 - 7 540
AK162441.1 - 7 490
BC019413.1 - 7 507 4166881
BC051924.1 - 7 567 5692787

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_031154, BC059130, FQ214048, J02744.

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
metabolic process
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
nitrobenzene metabolic process
xenobiotic catabolic process
negative regulation of ryanodine-sensitive calcium-release channel activity
positive regulation of ryanodine-sensitive calcium-release channel activity
cellular detoxification of nitrogen compound
cellular response to caffeine
glutathione transferase activity
receptor binding
transferase activity
enzyme binding
protein homodimerization activity
glutathione binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0