This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NDUFB2 SIRNA Assays ›
Gene Symbol:
Ndufb2
Gene Name:
NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 2
Gene Aliases:
1810011O01Rik, 8kDa, AGGG, AI325567, CI-AGGG
Chromosome Location:
Chr.6: 39592551 - 39599472 on Build GRCm38
UniGene ID:
Mm.29415
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s86384

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ndufb2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026612.3 NP_080888.1 2 211
GenBank mRNA AK007495.1 - 2 187
AK010642.1 - 2 218
AK019008.1 - 2 216
AK028334.1 - 220
AK216480.1 - 1 26
BC013510.1 - 2 221 4225177

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
oxidation-reduction process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0