This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CEP70 SIRNA Assays ›
Gene Symbol:
Cep70
Gene Name:
centrosomal protein 70
Gene Aliases:
6720484E09Rik, Bite, C030018L16Rik
Chromosome Location:
Chr.9: 99183595 - 99302065 on Build GRCm38
UniGene ID:
Mm.288739
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s86259

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cep70
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023873.3 NP_076362.2 3 213
GenBank mRNA AF242319.1 - 3 249
AK020174.1 - 3 551
AK020622.1 - 3 290
AK021095.1 - 3 344
AK049326.1 - 3 221
AK133048.1 - 3 219
BC050819.1 - 3 205 6517145
BC071260.1 - 3 213 6336351

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0