This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRPF18 SIRNA Assays ›
Gene Symbol:
Prpf18
Gene Name:
PRP18 pre-mRNA processing factor 18 homolog (yeast)
Gene Aliases:
2810441A10Rik
Chromosome Location:
Chr.2: 4622058 - 4652142 on Build GRCm38
UniGene ID:
Mm.38529
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s84697

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prpf18
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026045.3 NP_080321.2 9, 10 1062
GenBank mRNA AK028921.1 - 2, 3 199
AK035073.1 - 9, 10 1092
AK135707.1 - 9, 10 1102
AK135890.1 - 9, 10 1083
AK139720.1 - 9, 10 1050
AK161550.1 - 5, 6 506
BC004573.1 - 9, 10 1062 3586127
BC028306.1 - 9, 10 1065
BC058631.1 - 9, 10 1057 6402639

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_138523, AF244920.

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
potassium channel inhibitor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0