This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MBOAT2 SIRNA Assays ›
Gene Symbol:
Mboat2
Gene Name:
membrane bound O-acyltransferase domain containing 2
Gene Aliases:
2810049G06Rik, AU022889, AW547221, LPCAT4, Moact2, Oact2
Chromosome Location:
Chr.12: 24831487 - 24964400 on Build GRCm38
UniGene ID:
Mm.167671
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s84668

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mboat2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083341.1 NP_001076810.1 4, 5 465
NM_026037.3 NP_080313.2 5, 6 561
GenBank mRNA AB297383.1 - 5, 6 460
AB305046.1 - 5, 6 460
AK012931.1 - 4, 5 449
AK031824.1 - 4, 5 450
AK033106.1 - 2, 3 225
AK076045.1 - 4, 5 455
AK215458.1 - 3, 4 223
AU067514.1 - 5, 6 561
BC025020.1 - 4, 5 415 5364068

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
biological_process
phospholipid biosynthetic process
molecular_function
dihydrolipoamide branched chain acyltransferase activity
sterol O-acyltransferase activity
N-acetyltransferase activity
O-acyltransferase activity
palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
carnitine O-acyltransferase activity
acetyltransferase activity
C-acyltransferase activity
palmitoyltransferase activity
N-acyltransferase activity
acylglycerol O-acyltransferase activity
serine O-acyltransferase activity
O-acetyltransferase activity
O-octanoyltransferase activity
octanoyltransferase activity
O-palmitoyltransferase activity
S-acyltransferase activity
S-acetyltransferase activity
S-malonyltransferase activity
malonyltransferase activity
C-palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
succinyltransferase activity
N-succinyltransferase activity
O-succinyltransferase activity
S-succinyltransferase activity
sinapoyltransferase activity
O-sinapoyltransferase activity
peptidyl-lysine N6-myristoyltransferase activity
peptidyl-lysine N6-palmitoyltransferase activity
benzoyl acetate-CoA thiolase activity
3-hydroxybutyryl-CoA thiolase activity
3-ketopimelyl-CoA thiolase activity
N-palmitoyltransferase activity
myristoyltransferase activity
acyl-CoA N-acyltransferase activity
protein-cysteine S-myristoyltransferase activity
protein-cysteine S-acyltransferase activity
dihydrolipoamide S-acyltransferase activity
glucosaminyl-phosphotidylinositol O-acyltransferase activity
ergosterol O-acyltransferase activity
lanosterol O-acyltransferase activity
naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
2-methylhexanoyl-CoA C-acetyltransferase activity
butyryl-CoA 2-C-propionyltransferase activity
2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
L-2-aminoadipate N-acetyltransferase activity
UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
keto acid formate lyase activity
Ras palmitoyltransferase activity
azetidine-2-carboxylic acid acetyltransferase activity
1-acylglycerophosphocholine O-acyltransferase activity
peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
acetyl-CoA:L-lysine N6-acetyltransferase

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0