This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EXOSC7 SIRNA Assays ›
Gene Symbol:
Exosc7
Gene Name:
exosome component 7
Gene Aliases:
2610002K22Rik, AV212732, mKIAA0116
Chromosome Location:
Chr.9: 123113231 - 123136129 on Build GRCm38
UniGene ID:
Mm.41562
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s83281

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Exosc7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081188.1 NP_001074657.1 1 76
GenBank mRNA AK011294.1 - 1 63
AK085426.1 - 1 66
AK129057.1 - 1 28
AK217612.1 - 159
BC039655.1 - 1 61
BC052656.1 - 1 58 30041585
BC094932.1 - 1 69 30008071
BC132363.1 - 1 91 40130640
BC145675.1 - 1 91 40130978

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001100725, FQ226090, BF561514.

Gene Ontology Categories:

Function(s) Process(es)

rRNA processing
RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0